Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Individual Fungi Cultures

Shop All Carolina Biological Supply Products


Click to view available options
Media Type:
Potato Dextrose Agar
Rabbit Dung Agar
Packaging:
Plate
Tube
Vial
Scientific Name:
Aspergillus niger
Mucor hiemalis+
Penicillium notatum
Pilobolus crystallinus
Rhizopus stolonifer (+)
Storage Requirements:
20°C to 22°C
26°C
Description
Fungi are important decomposers, helping recycle organic materials in food webs, which makes them great subjects for ecological studies. Students can culture them for genetic studies. For studies in biochemistry, students can investigate the role yeast plays in the production of wine, beer and baked goods.
- Available in tube cultures, plate cultures or MicroKwik Cultures™
- Labeled by name and media on which they are cultivated
- Guaranteed for purity and genera
- Incubate at room temperature (26°C)
Carolina™ MicroKwick Cultlure™: Since a MicroKwik Culture can be refrigerated, you can order it ahead of time for use on short notice.
Specifications
Specifications
For Use With (Application) | Teach many topics in ecology, genetics, biochemistry and other disciplines. |
Packaging | Vial |
Storage Requirements | 26°C |
Quantity | Ea. |
Scientific Name | Penicillium notatum |
Includes | Carolina™ techniques for studying bacteria and fungi manual |
Sufficient For | Class of 30 students |
Media Type | Potato Dextrose Agar |
Product Type | Bacteria MicroKwik Culture™ |
Temperature (Metric) Range | 25°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction