Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Introductory Bacteria Sets
SDPSCI_ED

Catalog No. s07422nd Shop All Carolina Biological Supply Products
Click to view available options
Quantity:
3/Set
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • Bacillus subtilis (rods)
  • Micrococcus luteus (spheres)
  • Rhodospirillum rubrum (spirals)

Three bacteria tube cultures representing the common bacterial shapes of rods, spheres and spirals.

Specifications

Packaging Tube
Storage Requirements 20°C to 22°C
Quantity 3/Set
Includes Bacillus subtilis (rods), Micrococcus luteus (spheres) and Rhodospirillum rubrum (spirals)
Sufficient For Class of 30 students
Media Type Nutrient Agar
Product Type Introductory Bacteria Set
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.