Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Marine Dinoflagellate Mixture
SDPSCI_ED

Catalog No. s07477nd
Click to view available options
Quantity:
Ea.
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • Amphidinium
  • Gymnodinium
  • Prorocentrum
  • Materials for a class of 30 students

Many dinoflagellates are known to be photosynthetic, but a large fraction of these are in fact mixotrophic, combining photosynthesis with ingestion of prey.

  • Three representative dinoflagellates

Specifications

Storage Requirements 22°C
Quantity Ea.
Includes Three representative dinoflagellates Amphidinium, Gymnodinium, Prorocentrum
Sufficient For Class of 30 students
Product Type Marine Dinoflagellate Mixture
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.