Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Nutrient Agar, Prepared Media Bottles
SDPSCI_ED

Catalog No. p-4937416 Shop All Carolina Biological Supply Products
Click to view available options
Quantity:
125 mL
500 mL
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Catalog No. S21805ND

Supplier: Carolina Biological Supply 776360

Only null left
Add to Cart
Add to Cart
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Prepared Media Bottles - 100% Satisfaction Guaranteed!

The bottle is sealed to prevent contamination and drying. All bottles are glass, except where noted.

  • No autoclave needed
  • Free booklet on bottled media included
  • Note: C of A unavailable for this item

Specifications

Form Agar
Format Bottle
Manufacturing Origin United States
Media Type Nutrient Agar
Product Type Prepared Microbiology Media
Sterility Sterile
Target Organism Class Fastidious and Non-fastidious Organisms
Test Type Microbial Cultivation
Formulation Each glass bottle of bacteriological medium contains 125mL of ready-to-use material, except where noted. This amount is sufficient for pouring up to 5 plates or 15 to 20 tubes.
Quantity 125 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.