Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Painted Lady Butterfly 5-Larvae Culture
SDPSCI_ED

Catalog No. p-4655740
Click to view available options
Product Type:
Painted Lady Butterfly 5 Larvae Culture
Painted Lady Butterfly 5 Larvae Culture Coupon
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • 8 oz. cup with food
  • Five Vanessa cardui butterfly larvae
  • Larvae care instructions

Teach the concept of life cycle with live painted lady butterflies!

The larvae pupate and emerge as adult butterflies in as little as three weeks. Witnessing metamorphosis is an experience that students never forget!

  • At least three out of five larvae are guaranteed to emerge as healthy adult butterflies

Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.

Specifications

For Use With (Application) Carolina™ makes it easy to teach the concept of life cycle with live painted lady butterflies, witnessing metamorphosis is an experience that children never forget.
Storage Requirements 22°C to 25°C
Quantity 5/Container
Includes 8 oz. cup with food, five Vanessa cardui butterfly larvae, larvae care instructions
Sufficient For Class of 15 students
Product Type Painted Lady Butterfly 5 Larvae Culture Coupon
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.