Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Plant Kingdom Survey Set

Shop All Carolina Biological Supply Products


Click to view available options
:
Plant Kingdom Survey Set
Description
- Forty-seven specimens are labeled and mounted in twelve 6 x 5 in. glass front displays
- Includes Lichens, algae, fungi, liverworts, mosses, pteridophytes, gymnosperms, monocot angiosperms, dicot angiosperms, seed types, leaf venations, and parasitic plants
Specifications
Specifications
| Grade | K to 12 |
| Format | Set |
| Product Type | Plant Kingdom Survey Set Biorama |
| Includes | Lichens, algae, fungi, liverworts, mosses, pteridophytes, gymnosperms, monocot angiosperms, dicot angiosperms, seed types, leaf venations, and parasitic plants |
| Languages | English |
| Subject | Biology - Life Science |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction