Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Sporulating Moss

Shop All Carolina Biological Supply Products


Click to view available options
Quantity:
Ea.
Description
Includes
- Gametophyte with at least 12 sporophytes
- Genus shipped may vary seasonally
Specifications
Specifications
For Use With (Application) | Mosses are excellent terrarium subjects. |
Quantity | Ea. |
Sufficient For | Class of 30 students |
Media Type | A medium with a pH of 4.5 to 5.5 and low mineral content is preferable |
Product Type | Moss, Sporulating |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction