Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Termite Digestive Symbionts
SDPSCI_ED

Catalog No. S07669ND Shop All Carolina Biological Supply Products
Click to view available options
Quantity:
100/Container
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Catalog No. S07669ND

Supplier: Carolina Biological Supply 143740

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Trichonympha and Pyrsonympha, as well as other protozoa, live as symbionts in the termite's gut.

  • Shipped in live termites
  • With instructions

Specifications

For Use With (Application) Live as symbionts in the termite's gut.
Quantity 100/Container
Includes Instructions
Sufficient For Class of 60 students
Product Type Termite Digestive Symbiont
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.