Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caronte Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547055
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547055 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547055 Supplier Invitrogen™ Supplier No. PA547055
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) Cerberus-1 is observed and less than 1% cross-reactivity with rhDAN and recombinant mouse Gremlin is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

Chicken Caronte, the ortholog of mammalian Cerberus, is a member of the Cerberus/DAN family of secreted glycoproteins. There are at least 5 Dan Domain (DAND) family members including DAN (DAND1), Gremlin/Drm (DAND2), PRDC (Protein Related to Dan and Cerberus; DAND3), Cerberus (DAND4), and Coco (DAND5). Caronte is expressed in the left paraxial mesoderm and is the molecule responsible for transducing left-sided positional information from the node to the periphery of the lateral plate mesoderm (LPM) during chicken embryonic development. Caronte functions as a BMP antagonist, inducing Nodal and downstream genes in the LPM by interfering with the repressive activity of bilaterally-produced BMPs. In addition, antagonism of BMPs by Caronte induces the expression of the Lefty gene in the midline.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caronte
Applications Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Alias CER1; Cerberus; cerberus 1, cysteine knot superfamily, homolog; cerberus 1, DAN family BMP antagonist; cerberus-related 1; cerberus-related protein; Chicken Caronte; DAN domain family member 4; DAND4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant chicken Caronte Asp16-Asn272.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chicken
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.