Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
β-Carotene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
1 g
3 x 100 mg
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications
Specifications
CAS | 7235-40-7 |
Physical Form | Neat |
Linear Formula | C40H56 |
MDL Number | MFCD00001556 |
Synonym | beta,beta-Carotene; Provitamin A |
Type | Certified Reference Material |
Quantity | 1 g |
Formula Weight | 536.87 |
Grade | Certified Reference Material |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction