Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Carrying Bag
SDPSCI_ED

Catalog No. S00285 Shop All 3B Scientific Products
Click to view available options
:
Carrying Bag; Color: Blue
This item is not returnable. View return policy
This item is not returnable. View return policy

Carrying Bag for patient care manikins.

  • For use with:
    • Simple Simon Patient Care Manikin (W45070)
    • Simple Susie Patient Care Simulator (W45057)
    • Susie Simon Patient Care Manikin with Ostomy (W45011)
  • Dimensions: 35.4 x 14.6 x 11.0 in.
  • Weight: 2.98 lb.

Specifications

Product Type Carrying Bag
Color Blue
For Use With (Application) Carrying Patient Care Manikins
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.