Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARS Antibody (8B11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CARS |
---|---|
Clone | 8B11 |
Dilution | Western Blot 1:500-1:2000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
CARS Monoclonal antibody specifically detects CARS in Human samples. It is validated for Western Blot, ELISASpecifications
CARS | |
Western Blot 1:500-1:2000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CARS1, CysRS, cysteine translase, cysteine tRNA ligase 1, cytoplasmic, Cysteine--tRNA ligase, cysteinyl-tRNA synthetase, cysteinyl-tRNA synthetase, cytoplasmic, EC 6.1.1, EC 6.1.1.16, MGC:11246 | |
A synthesized peptide derived from Human CARS [UniProt P49589] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
8B11 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
833 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title