Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARS Antibody (CL2309), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CARS |
---|---|
Clone | CL2309 |
Dilution | Immunohistochemistry 1:2500-1:5000 |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Description
CARS Monoclonal specifically detects CARS in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CARS | |
Immunohistochemistry 1:2500-1:5000 | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CARS1, CysRS, cysteine translase, cysteine tRNA ligase 1, cytoplasmic, Cysteine--tRNA ligase, cysteinyl-tRNA synthetase, cysteinyl-tRNA synthetase, cytoplasmic, EC 6.1.1, EC 6.1.1.16, MGC:11246 | |
CARS | |
IgG2b | |
Protein A purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL2309 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
A8MVQ3 | |
833 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SQCNLYMAARKAVRKRPNQALLENIALYLTHMLKIFGAVEEDSSLGFPVGGPGTSLSLEATVMPYLQVLSEFREGVRKIAREQKVPEILQLSDALRDNILPELGVRFEDHEGLPTVVKLVDRNTLLKEREEKRRVE | |
Primary | |
Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title