Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CARS Monoclonal Antibody (CL2302)
GREENER_CHOICE

Catalog No. PIMA524634
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524634 Supplier Invitrogen™ Supplier No. MA524634
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: SQCNLYMAAR KAVRKRPNQA LLENIALYLT HMLKIFGAVE EDSSLGFPVG GPGTSLSLEA TVMPYLQVLS EFREGVRKIA REQKVPEILQ LSDALRDNIL PELGVRFEDH EGLPTVVKLV DRNTLLKERE EKRRVE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%. Binds to an epitope located within the peptide sequence RPNQALLENIALYLT as determined by overlapping synthetic peptides.

This gene encodes a class 1 aminoacyl-tRNA synthetase, cysteinyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. This gene is one of several located near the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL2302
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CARS1
Gene Accession No. P49589
Gene Alias CA3; CARS; cars protein; CARS1; CYSRS; cysteine translase; cysteine tRNA ligase 1, cytoplasmic; cysteine--tRNA ligase, cytoplasmic; Cysteinyl-tRNA synthetase; cysteinyl-tRNA synthetase 1; cysteinyl-tRNA synthetase, cytoplasmic; LOW QUALITY PROTEIN: cysteine--tRNA ligase, cytoplasmic; MGC:11246; sb:cb71
Gene Symbols CARS, CARS1
Host Species Mouse
Immunogen Recombinant protein fragment of Human CARS (Product #RP-101867)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 833
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.