Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CARS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597539
Description
CARS Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a class 1 aminoacyl-tRNA synthetase, cysteinyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. This gene is one of several located near the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms.
Specifications
CARS | |
Polyclonal | |
Unconjugated | |
CARS1 | |
CA3; CARS; cars protein; CARS1; CYSRS; cysteine translase; cysteine tRNA ligase 1, cytoplasmic; cysteine--tRNA ligase, cytoplasmic; Cysteinyl-tRNA synthetase; cysteinyl-tRNA synthetase 1; cysteinyl-tRNA synthetase, cytoplasmic; LOW QUALITY PROTEIN: cysteine--tRNA ligase, cytoplasmic; MGC:11246; sb:cb71 | |
Rabbit | |
Protein G | |
RUO | |
27267, 833 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P49589, Q9ER72 | |
CARS, CARS1 | |
Recombinant Human Cysteine--tRNA ligase, cytoplasmic protein (201-500AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction