Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CARS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106960
Description
Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 85%; Rabbit: 86%; Rat: 85%.
CARS is a class 2 aminoacyl-tRNA synthetase, cysteinyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. It catalyzes the chemical reaction: ATP + L-cysteine + tRNA(Cys) = AMP + diphosphate + L-cysteinyl-tRNA(Cys).
Specifications
CARS2 | |
Polyclonal | |
Unconjugated | |
CARS2 | |
CARS2; COXPD27; cysRS; cysteine tRNA ligase 2, mitochondrial (putative); Cysteinyl-tRNA synthetase; cysteinyl-tRNA synthetase 2, mitochondrial (putative); OK/SW-cl.10; Probable cysteine--tRNA ligase, mitochondrial | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79587 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
Q9HA77 | |
CARS2 | |
Synthetic peptide directed towards the N-terminal region of Human CARS2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction