Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Carteolol hydrochloride

Catalog No. p-7214489
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51781-21-6
Molecular Formula C16H25ClN2O3
Molecular Weight (g/mol) 328.84
MDL Number MFCD00941499
InChI Key FYBXRCFPOTXTJF-UHFFFAOYNA-N
IUPAC Name hydrogen 5-[3-(tert-butylamino)-2-hydroxypropoxy]-1,2,3,4-tetrahydroquinolin-2-one chloride
SMILES [H+].[Cl-].CC(C)(C)NCC(O)COC1=CC=CC2=C1CCC(=O)N2

Specifications

Melting Point 278.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: soluble. Other solubilities: moderately soluble in methanol. slightly soluble, in acetic acid. insoluble in chloroform, ether,, dichloromethane
Formula Weight 328.83
Chemical Name or Material Carteolol hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes serious eye irritation.

Suspected of damaging the unborn child.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

If eye irritation persists: Get medical advice/attention.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 257-415-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.