Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Carvone, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111013585
Click to view available options
Quantity:
1 mL
25 mL
100 mL
Featured Industries: Agriculture and Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 2244-16-8
Density 0.960 g/mL (at 20°C)
Flash Point 89°C - Closed Cup
Physical Form Neat
Linear Formula C10H14 O
MDL Number MFCD00062997
Boiling Point 228°C to 230°C
Synonym (+)-p-Mentha-6,8-diene 2-one; (S)-5-Isopropenyl-2-methyl-2-cyclohexenone
Type Analytical Standard
Quantity 100 mL
Formula Weight 150.22
Percent Purity ≥98.5% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.