Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586130
Description
Purity is > 95% by SDS-PAGE.
Proteins that carry a nuclear localization signal (NLS) are transported into the nucleus by the importin-alpha/beta heterodimer. Importin-alpha binds the NLS, while importin-beta mediates translocation through the nuclear pore complex. After translocation, RanGTP binds importin-beta and displaces importin-alpha. Importin-alpha must then be returned to the cytoplasm, leaving the NLS protein behind. The protein encoded by this gene binds strongly to NLS-free importin-alpha, and this binding is released in the cytoplasm by the combined action of RANBP1 and RANGAP1. In addition, the encoded protein may play a role both in apoptosis and in cell proliferation.
Specifications
CAS | |
Polyclonal | |
Unconjugated | |
CSE1L | |
2610100P18Rik; AA407533; Capts; CAS; Cellular apoptosis susceptibility protein; chromosome segregation 1 like; chromosome segregation 1-like (S. cerevisiae); chromosome segregation 1-like protein; CSE1; CSE1 chromosome segregation 1 like; CSE1 chromosome segregation 1-like; CSE1 chromosome segregation 1-like (yeast); CSE1L; Exp2; exportin-2; Importin-alpha re-exporter; MGC117283; MGC130036; MGC130037; XPO2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
110750, 1434, 362273 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P55060, Q9ERK4 | |
CSE1L | |
Synthetic peptide, corresponding to amino acids N-terminus of Human CSE1L. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction