Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAS Recombinant Rabbit Monoclonal Antibody (JU34-33)
GREENER_CHOICE

Catalog No. PIMA534638
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534638 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534638 Supplier Invitrogen™ Supplier No. MA534638
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Mouse testis tissue, SiHa, SK-BR-3, PC-3M, LOVO, SH-SY5Y, human colon cancer tissue, LOVO.

Proteins that carry a nuclear localization signal (NLS) are transported into the nucleus by the importin-alpha/beta heterodimer. Importin-alpha binds the NLS, while importin-beta mediates translocation through the nuclear pore complex. After translocation, RanGTP binds importin-beta and displaces importin-alpha. Importin-alpha must then be returned to the cytoplasm, leaving the NLS protein behind. The protein encoded by this gene binds strongly to NLS-free importin-alpha, and this binding is released in the cytoplasm by the combined action of RANBP1 and RANGAP1. In addition, the encoded protein may play a role both in apoptosis and in cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAS
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JU34-33
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CSE1L
Gene Accession No. P55060
Gene Alias 2610100P18Rik; AA407533; Capts; CAS; Cellular apoptosis susceptibility protein; chromosome segregation 1 like; chromosome segregation 1-like (S. cerevisiae); chromosome segregation 1-like protein; CSE1; CSE1 chromosome segregation 1 like; CSE1 chromosome segregation 1-like; CSE1 chromosome segregation 1-like (yeast); CSE1L; Exp2; exportin-2; Importin-alpha re-exporter; MGC117283; MGC130036; MGC130037; XPO2
Gene Symbols CSE1L
Host Species Rabbit
Immunogen Recombinant protein within Human Cellular Apoptosis Susceptibility aa 860-971.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1434
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.