Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cas12j Polyclonal Antibody

Catalog No. PIPA5143500
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143500 500 μL
PIPA5143597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5143500 Supplier Invitrogen™ Supplier No. PA5143500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cas12j Polyclonal Antibody for Western Blot, ICC/IF

There has been much recent interest in gene editing and other genetic manipulations by CRISPR-Cas family enzymes. The lab of Nobel prize winner Jennifer Doudna characterized these Cas enzymes and showed that they could be used to perform CRISPR, but had the significant advantage that they were much smaller in molecular size than other Cas family enzymes such as the widely used Cas9 enzymes from S. pyogenes and S. aureus. One of these enzymes is referred to as Cas theta and also known as Cas-12j. The use of such smaller Cas family enzymes leaves more room for other nucleic acid sequences in AAV and other viral vectors which typically have a limited DNA capacity, thus allowing more versatility for new CRISPR based manipulations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cas12j
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5 mM sodium azide
Gene Alias Cas theta
Host Species Rabbit
Immunogen Full length Cas12j expressed in and purified from E. coli.
Purification Method Affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.