Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cas9, D10A/H840A Mutant, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABC1490MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABC1490MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC1490MI Supplier MilliporeSigma™ Supplier No. ABC1490
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRISPR-associated endonuclease Cas9/Csn1 (UniProt Q99ZW2; also known as SpyCas9) is encoded by the Streptococcus pyogenes serotype M1 gene Cas9 (also known as csn1) gene (Gene ID 901176). Clustered regularly interspaced short palindromic repeats (CRISPRs) are DNA loci containing short repetitions of base sequences complementary to antecedent mobile elements and target invading nucleic acids. Each repetition is followed by short segments of spacer DNA from previous exposures to a virus. Cas9 is an essential endonuclease in Streptococcus pyogenes serotype M1's CRISPR immune system that confers resistance to foreign genetic elements such as plasmids and phages. CRISPR clusters are transcribed and processed into CRISPR RNA (crRNA). In type II CRISPR systems, correct processing of pre-crRNA requires a trans-encoded small RNA (tracrRNA), endogenous ribonuclease 3 (rnc), and Cas9. Cas9/crRNA/tracrRNA endonucleolytically cleaves linear or circular dsDNA target complementary to the spacer. The target strand not complementary to crRNA is first cut endonucleolytically, and then trimmed 3&prime-5&prime exonucleolytically. Mutations of the catalytic residues (D10A in RuvC and H840A in HNH) convert Cas9 into DNA nickases (Cas9n) that cleave only the strand complementary to the crRNA, minimizing off-target activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cas9, D10A/H840A Mutant
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene cas9, csn1
Gene Accession No. Q99ZW2
Host Species Rabbit
Immunogen A mixture of two KLH-conjugated peptides corresponding to the Cas9 target sites sequences with D10A and H840A mutations.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Secondary & Control Antibodies
Primary or Secondary Primary
Gene ID (Entrez) AKA60242.1
Test Specificity This polyclonal antibody will detect Cas9 with D10A mutation in the RuvC-I region and/or H840A mutation in the HNH domain (e.g. AKA60242.1), but not Cas9 with neither D10A nor H840A mutation (e.g. NP_269215).
Target Species All species
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.