Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cas9 Monoclonal Antibody (8C1-F10), Active Motif™
SDP

Catalog No. 502277258
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7258 50 μg
50-199-3676 10 μg
50-199-3675 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 50-227-7258 Supplier Active Motif Supplier No. 61957
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cas9 is a RNA-guided DNA endonuclease enzyme associated with the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) adaptive immunity system in bacteria and genetic engineering. Staphylococcus aureus derived Cas9 has a number of properties that make it advantageous for genome editing, including its small size, high efficiency, nickase activity, and apparent specificity. SaCas9 principally recognizes an NNGRRT protospacer adjacent motif (PAM) and cleaves target DNA at rates comparable to SpCas9 (Cas9 derived from S. pyogenes) with an ideal spacer lengths of 24 to 20 nucleotides. In comparison to SpCas9, SaCas9 is smaller (1368 AA versus 1053 AA) and generates indels at a comparable rate when directed against target DNA with the mutually permissive NGGRRT PAM. This makes it the preferred enzyme for all-in-one delivery of Cas9 and multiple gRNA expression cassettes with Adeno-associated vectors.

Specifications

Antigen Cas9
Applications ChIP Assay, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8C1-F10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene CASP9
Gene Accession No. P55211, Q8C3Q9
Gene Alias APAF3; APAF-3; apoptotic protease activating factor 3; Apoptotic protease Mch-6; Apoptotic protease-activating factor 3; cas9; CASP 9; CASP9; CASP-9; caspase 9; caspase 9, apoptosis-related cysteine peptidase; Caspase9; Caspase-9; Caspase-9 precursor; Caspase-9 subunit p10; Caspase-9 subunit p35; CASPASE-9c; CRISPR-Cas9; CTD; ICE-LAP6; ICE-like apoptotic protease 6; MCH6; OTTHUMP00000002324; PPP1R56; protein phosphatase 1, regulatory subunit 56; RNCASP9; RP11-265F14.3
Gene Symbols CASP9
Host Species Mouse
Immunogen This antibody was raised against a recombinant protein within the N-terminal region of Streptococcus pyogene Cas9. This antibody should recognize Cas9 and dCas9 based on the antigen design.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12371, 842
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.