Learn More
Cas9 Monoclonal Antibody (OTI6D9), TrueMAB™, OriGene

Description
Specifications
Specifications
| Antigen | CAS9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI6D9 |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene Alias | Cas9Cas9; Cas9 |
| Host Species | Mouse |
| Immunogen | Human recombit protein fragment corresponding to amino acids 1-1166 of human CAS9 produced in E.coli. |
| Purification Method | Affinity Chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.