Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cas9 Mouse anti-Bacteria, All, Clone: 5F1, MilliporeSigma™

Catalog No. MAC141MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAC141MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAC141MI Supplier MilliporeSigma Supplier No. MAC141
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Cas-9, C-terminal, clone 5F1 is a mouse monoclonal antibody that detects Cas-9 in Staphylococcus aureus and its identity has been confimed by Isotyping Test and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cas9
Applications Western Blot
Classification Monoclonal
Clone 5F1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Cas9
Host Species Mouse
Immunogen His-tagged recombinant Cas-9 from Staphylococcus aureus.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Secondary and Control Antibodies
Primary or Secondary Primary
Test Specificity Clone 5F1 detects Cas-9 in Staphylococcus aureus.
Target Species Bacteria, All species
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.