Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cas9 Polyclonal Antibody, GenCRISPR™, GenScript™

Catalog No. 502120639
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0639 Supplier Genscript Corporation Supplier No. A01885100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRISPR/Cas9 system is one powerful gene editing tool to specifically modify target genes. Cas9 nuclease is an RNA-guided endonuclease that can catalyze cleavage of double stranded DNA. The GenCrispr Cas9 Antibody is a rabbit polyclonal antibody with Streptococcus pyogene (S. pyogenes) Cas9 protein as the immunization antigen. This antibody is used to detect the expression level of CRISPR/Cas9 in target cells.

Cas9 Polyclonal antibody specifically detects Cas9 in Bacteria samples. It is validated for Western Blot

Specifications

Antigen Cas9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol and no preservative; pH 7.4
Gene Alias cas9; CASP 9; Caspase9; Caspase-9 precursor; CASPASE-9c; CRISPR9; CRISPR-Cas9; CTD; OTTHUMP00000002324; RNCASP9; RP11-265F14.3; s aureus
Host Species Rabbit
Immunogen Recombinant Streptococcus pyogene (S. pyogenes) Cas9 protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.