Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cas9, Rabbit anti-Bacteria, Polyclonal, Millipore Sigma™

Catalog No. ABS220225UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS220225UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS220225UG Supplier MilliporeSigma Supplier No. ABS220225UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Cas9 Antibody, Cat. No. ABS2202, is a highly specific rabbit polyclonal antibody that targets Cas9 and has been tested for use in Immunocytochemistry and Western Blotting.

CRISPR-associated endonuclease Cas9/Csn1 (UniProt: Q99ZW2; also known as SpCas9, SpyCas9) is encoded by the Cas9 (also known as Csn1) gene (Gene ID: 901176) in Streptococcus pyogenes. CRISPR (clustered regularly interspaced short palindromic repeat) is an adaptive immune system that provides protection against mobile genetic elements (viruses, transposable elements and conjugative plasmids). CRISPR clusters contain spacers, sequences complementary to antecedent mobile elements, and target invading nucleic acids. Cas9 has a bi-lobed architecture with a recognition lobe (residues 41-425) and a discontinuous nuclease lobe (NUC, residues 1-40 and 453-1053); the crRNA-target DNA lies in a channel between the 2 lobes. The NUC lobe has 2 endonuclease domains. The discontinuous RuvC-like domain in NUC cleaves the target DNA non-complementary to crRNA while the HNH nuclease domain cleaves the target DNA complementary to crRNA. Cas9 serves as a RNA-guided DNA endonuclease that cleaves double-stranded DNA targets with a protospacer adjacent motif (PAM) and complementarity to the guide RNA. The homologous region of the Staphylococcus aureus is not closely related in amino acid sequence in Streptococcus pyogenes, hence this antibody does not recognize that protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cas9
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Cas9 in HEK293 cells transfected with Cas9 from Streptococcus pyogenes.Immunocytochemistry Analysis: 1μg/mL from a representative lot detected Cas9 in non-transfected HEK293H and HEK293H transfected with pCDNA3.0-Cas9.Western Blotting Analysis: 10μg/mL from a representative lot detected Cas9 in HEK293H cell lysate from cells transfected with pCDNA3.0-Cas9.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias SpCas9;CRISPR-associated endonuclease Cas9
Gene Symbols Cas9;Csn1
Host Species Rabbit
Immunogen KLH-conjugated linenar peptide corresponding to 18 amino acids from the C-terminal region of Streptococcus pyogenes.
Molecular Weight of Antigen 158.44 kDa calculated.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Cas9 in Streptococcus pyogenes. It targets an epitope within 18 amino acids from the C-terminal region. This antibody does not react with Cas9 from Staphylococcus aureus.
Target Species Bacteria
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.