Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cas9 Rabbit anti-Bacteria, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. AB35648025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB356480-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB356480-25 Supplier MilliporeSigma Supplier No. AB35648025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Cas9 (Rabbit Polyclonal), Cat. No. AB356480, is a highly specific rabbit polyclonal antibody that targets CRISPR-associated endonuclease Cas9 and has been tested in Immunofluorescence and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cas9
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide.
Gene Alias SaCas9;CRISPR-associated endonuclease Cas9
Gene Symbols Cas9
Host Species Rabbit
Immunogen His-tagged recombinant fragment corresponding to 251 amino acids from the C-terminal end of CRISPR-associated endonuclease Cas9 from Staphylococcus aureus.
Molecular Weight of Antigen ∽55 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Cas9 from Staphylococcus aureus. It targets an epitope within 251 amino acids from the C-terminal end. It does not react with Cas9 from Streptococcus pyogenes.
Target Species Bacteria
Content And Storage Stable for 1 year at -20°C from date of receipt.
Product Type Antibodies
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.