Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Casamino Acids

Catalog No. p-7112260 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
227 g
454 g

Supplemented to microbiological growth media.

Applications: Casamino acids is supplemented to microbiological growth media.
Product Description: Casamino acids are a mixture of amino acids (except tryptophan) obtained from acid hydrolysis of casein.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature (15-30 °C).
Form Powder
Product Type Media Supplement
Quantity 227 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.