Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CASC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561989 Supplier Invitrogen™ Supplier No. PA561989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSHINHNGNP GTSKQNPSSP LQRLIPGSNL DSEPRIQTDI LKQATKDRVS DFHKLKQSRF FDENESPVDP QHGSKLADYN GDDG Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.

The increased expression level of this gene is associated with HER-2/neu proto-oncogene overexpression. Amplification and resulting overexpression of this proto-oncogene are found in approximately 30% of human breast and 20% of human ovarian cancers. Further characterization of the product of this gene may yield insight into the fundamental biology and pathogenetic effects of HER-2/neu overexpression in human breast and ovarian cancer cells. Alternatively spliced variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CASC4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CASC4
Gene Accession No. Q6P4E1
Gene Alias cancer susceptibility candidate 4; cancer susceptibility candidate gene 4 protein; Cancer susceptibility candidate gene 4 protein homolog; Casc4; D130060C09Rik; gene associated with HER-2/neu overexpression; H63; Protein CASC4; UNQ2573/PRO6308; VGFG2573
Gene Symbols CASC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CASC4. Recombinant protein control fragment (Product #RP-99876).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113201
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.