Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CASC5 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002586AF647
Description
CASC5 Polyclonal antibody specifically detects CASC5 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
CASC5 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
AF15Q14, Blinkin, blinkin, bub-linking kinetochore protein, Bub-linking kinetochore protein, cancer susceptibility candidate 5, Cancer susceptibility candidate gene 5 protein, Cancer/testis antigen 29, CT29KIAA1570, D40, hKNL-1, hSpc105, kinetochore null 1 homolog, Kinetochore-null protein 1, KNL1, KNL1ALL1-fused gene from chromosome 15q14 protein, protein CASC5, Protein D40/AF15q14 | |
The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human Cancer Susceptibility Candidate 5 using the numbering given in entry NP_733468.2 | |
0.1 mL | |
DNA Repair | |
57082 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction