Learn More
Invitrogen™ Casc5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588989
Description
Immunogen sequence: MDGVSSEANE ENDNIERPVR RRHSSILKPP RSPLQDLRGG NERVQESNAL RNKKNSRRVS FADTIKVFQT ESHMKIVRKS EMEGCSAMVP SQLQLLPPGF KRFSCLSLPE TETGENLLLI QNKKLEDNYC EITGMNTLLS APIHTQMQQK EFSIIEHTRE RKHANDQTVI FSDENQMDLT SSHTVMITKG LLDNPISEKS TKIDTTSFLA NLKLHTEDSR MKKEVNFSVD QNTSSENKID FNDFIKRLKT GKCSAFPDVP DKENFEIPIY SKEPNSASST HQMHVSLKED ENNSNITRLF; Positive Samples: HepG2, U-87MG, HeLa, Jurkat; Cellular Location: Chromosome, Nucleus, centromere, kinetochore.
Performs two crucial functions during mitosis: it is essential for spindle-assembly checkpoint signaling and for correct chromosome alignment. Required for attachment of the kinetochores to the spindle microtubules. Directly links BUB1 and BUB1B to kinetochores. Part of the MIS12 complex, which may be fundamental for kinetochore formation and proper chromosome segregation during mitosis. Acts in coordination with CENPK to recruit the NDC80 complex to the outer kinetochore.
Specifications
Casc5 | |
Polyclonal | |
Unconjugated | |
KNL1 | |
2310043D08Rik; 5730505K17Rik; AF15Q14; ALL1-fused gene from chromosome 15q14 protein; Blinkin; blinkin, bub-linking kinetochore protein; Bub-linking kinetochore protein; cancer susceptibility candidate 5; cancer susceptibility candidate gene 5 protein; Cancer/testis antigen 29; CASC5; CT29; D40; hKNL-1; hSpc105; KIAA1570; kinetochore null 1 homolog; kinetochore scaffold 1; Kinetochore-null protein 1; KNL1; MCPH4; microcephaly, primary autosomal recessive 4; PPP1R55; Protein CASC5; Protein D40/AF15q14; protein phosphatase 1, regulatory subunit 55; Rad51; RAD51 homolog; Spc7 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
311327, 57082 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.69 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8NG31 | |
KNL1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CASC5 (NP_7334683). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.