Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Casc5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565573
Description
Immunogen sequence: DVTKQVIQTHV NAGEAPDPVI TSNVPCFHSI KPNLNNLNGK TGEFLAFQTV HLPPLPEQLL ELGNKAHNDM HIVQATEIHN INIISSNAKD SRDEENKKSH NGAETTSLPP KTVFKDKVRR CSLGIFLPRL PNKRNCSVTG IDDLEQ Highest antigen sequence identity to the following orthologs - mouse 60%, rat 60%.
Performs two crucial functions during mitosis: it is essential for spindle-assembly checkpoint signaling and for correct chromosome alignment. Required for attachment of the kinetochores to the spindle microtubules. Directly links BUB1 and BUB1B to kinetochores. Part of the MIS12 complex, which may be fundamental for kinetochore formation and proper chromosome segregation during mitosis. Acts in coordination with CENPK to recruit the NDC80 complex to the outer kinetochore.
Specifications
Casc5 | |
Polyclonal | |
Unconjugated | |
KNL1 | |
2310043D08Rik; 5730505K17Rik; AF15Q14; ALL1-fused gene from chromosome 15q14 protein; Blinkin; blinkin, bub-linking kinetochore protein; Bub-linking kinetochore protein; cancer susceptibility candidate 5; cancer susceptibility candidate gene 5 protein; Cancer/testis antigen 29; CASC5; CT29; D40; hKNL-1; hSpc105; KIAA1570; kinetochore null 1 homolog; kinetochore scaffold 1; Kinetochore-null protein 1; KNL1; MCPH4; microcephaly, primary autosomal recessive 4; PPP1R55; Protein CASC5; Protein D40/AF15q14; protein phosphatase 1, regulatory subunit 55; Rad51; RAD51 homolog; Spc7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57082 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NG31 | |
KNL1 | |
Recombinant Human Casc5. Recombinant protein control fragment (Product #RP-106240). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction