Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASC5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986936
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6936 Supplier Bioss Supplier No. BS5959R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Performs two crucial functions during mitosis: it is essential for spindle-assembly checkpoint signaling and for correct chromosome alignment. Required for attachment of the kinetochores to the spindle microtubules. Directly links BUB1 and BUB1B to kinetochores. Part of the MIS12 complex, which may be fundamental for kinetochore formation and proper chromosome segregation during mitosis. Acts in coordination with CENPK to recruit the NDC80 complex to the outer kinetochore.

Specifications

Antigen CASC5
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KNL1
Gene Accession No. Q8NG31
Gene Alias 2310043D08Rik; 5730505K17Rik; AF15q14; ALL1-fused gene from chromosome 15q14 protein; Blinkin; blinkin, bub-linking kinetochore protein; Bub-linking kinetochore protein; cancer susceptibility candidate 5; Cancer susceptibility candidate gene 5 protein; Cancer/testis antigen 29; CASC5; CT29; D40; hKNL-1; hSpc105; KIAA1570; kinetochore null 1 homolog; Kinetochore scaffold 1; Kinetochore-null protein 1; KNL1; MCPH4; microcephaly, primary autosomal recessive 4; PPP1R55; Protein CASC5; Protein D40/AF15q14; protein phosphatase 1, regulatory subunit 55; Rad51; RAD51 homolog; Spc7
Gene Symbols KNL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CASC5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57082
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.