Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CASD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569624
Description
This target displays homology in the following species: Cow: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
O-acetyltransferase that catalyzes 9-O-acetylation of sialic acids (PubMed:20947662, PubMed:26169044). Sialic acids are sugars at the reducing end of glycoproteins and glycolipids, and are involved in various processes such as cell-cell interactions, host-pathogen recognition (PubMed:20947662, PubMed:26169044).
Specifications
CASD1 | |
Polyclonal | |
Unconjugated | |
CASD1 | |
C7orf12; Cas1; CAS1 domain containing 1; CAS1 domain-containing protein 1; Casd1; Cast1; N-acetylneuraminate 9-O-acetyltransferase; Nbla04196; O-acetyltransferase; Sialate O-acetyltransferase; SOAT; WUGSC:H_GS542D18.2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
64921 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96PB1 | |
CASD1 | |
synthetic peptide directed towards the N terminal region of human CASD1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction