Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CASD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560700 Supplier Invitrogen™ Supplier No. PA560700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SIAKPHVIVA GAATWSIKIH NGSSEALSQY KMNITSIAPL LEKLAKTSDV YWVLQDPVYE DLLSENRKMI TNEKIDAY Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

O-acetyltransferase that catalyzes 9-O-acetylation of sialic acids (PubMed:20947662, PubMed:26169044). Sialic acids are sugars at the reducing end of glycoproteins and glycolipids, and are involved in various processes such as cell-cell interactions, host-pathogen recognition (PubMed:20947662, PubMed:26169044).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CASD1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CASD1
Gene Accession No. Q96PB1
Gene Alias C7orf12; Cas1; CAS1 domain containing 1; CAS1 domain-containing protein 1; Casd1; Cast1; N-acetylneuraminate 9-O-acetyltransferase; Nbla04196; O-acetyltransferase; Sialate O-acetyltransferase; SOAT; WUGSC:H_GS542D18.2
Gene Symbols CASD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CASD1. Recombinant protein control fragment (Product #RP-92709).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64921
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.