Learn More
Description
- Powder
- CAS: 9000-71-9
This product is an Educational Grade Chemical — no C of A is available.
Specifications
Specifications
| For Use With (Application) | Laboratory Chemicals |
| Name | Casein |
| Quantity | 100 g |
| Packaging | Solid |
| Form | Solid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
