Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Casein Antibody (SR1341), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Casein |
---|---|
Clone | SR1341 |
Dilution | Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
Casein Monoclonal antibody specifically detects Casein in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Casein | |
Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
alpha, alpha-S1-casein, CASA, casein alpha s1, MGC149368 | |
A synthesized peptide derived from human Casein (Uniprot #: P47710) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1341 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cell Biology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1446 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title