Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein, Hammarsten Grade

Catalog No. aaj1284022
Click to view available options
Quantity:
1 lb.
100 g
11.3 kg (25 lb.)
2.2 kg (5 lb.)

Cream to light yellow, lyophilized powder. | 75000 to 100000 g/mol

  • Cream to light yellow
  • Protein: ≥95%
  • Melting point: ∽280°C (dec.)
  • MDLNumber: MFCD00081481
  • CAS: 9000-71-9
TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 75000 to 100000
Name Casein
Quantity 100 g
Packaging Plastic bottle
Sensitivity Hygroscopic
Form Lyophilized

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.