Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Casein, Hammarsten Grade
Click to view available options
Quantity:
1 lb.
100 g
11.3 kg (25 lb.)
2.2 kg (5 lb.)
Description
- Cream to light yellow
- Protein: ≥95%
- Melting point: ∽280°C (dec.)
- MDLNumber: MFCD00081481
- CAS: 9000-71-9
Specifications
Specifications
| Molecular Weight (g/mol) | 75000 to 100000 |
| Quantity | 1 lb. |
| Packaging | Plastic Bottle |
| Product Type | Casein |
| Content And Storage | Ambient Temperatures, Store under Nitrogen |
| Sensitivity | Hygroscopic |
| Form | Lyophilized |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction