Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein, Hammarsten Grade

Catalog No. AAJ12840P5
Change view
Click to view available options
Quantity:
1 lb.
100 g
11.3 kg (25 lb.)
2.2 kg (5 lb.)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ12840P5 2.2 kg (5 lb.)
AAJ1284022 100 g
AAJ12840P2 1 lb.
AAJ12840Q1 11.3 kg (25 lb.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ12840P5 Supplier Thermo Scientific Supplier No. J12840P5
Only null left
Add to Cart

Cream to light yellow, lyophilized powder. | 75000 to 100000 g/mol

  • Cream to light yellow
  • Protein: ≥95%
  • Melting point: ∽280°C (dec.)
  • MDLNumber: MFCD00081481
  • CAS: 9000-71-9
TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 75000 to 100000
Name Casein
Quantity 2.2 kg (5 lb.)
Packaging Plastic bottle
Sensitivity Hygroscopic
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.