Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein, High Nitrogen

Catalog No. aaj12845q1
Click to view available options
Quantity:
11.3 kg (25 lb.)
2.2 kg (5 lb.)

Prepared from New Zealand casein exclusively.

  • Format: White to off-white powder
  • Protein (Dry basis): ≥95.0%
  • Melting point: ∽280°C (dec.)
  • MDLNumber: MFCD00081481
  • CAS: 9000-71-9
TRUSTED_SUSTAINABILITY

Specifications

Name Casein
Quantity 11.3 kg (25 lb.)
Packaging Plastic bottle
Sensitivity Hygroscopic
Form Powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.