Learn More
Casein (kappa) Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 600830

Description
Positive control: Mouse mammary gland Cellular location: milk.
Casein (kappa) Polyclonal antibody specifically detects Casein (kappa) in Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
Casein (kappa) | |
Polyclonal | |
Unconjugated | |
CSN3 | |
casein kappa; CASK; CNS10; CSN10; CSN3; CSNK; kappa-casein; KCA | |
A synthetic peptide corresponding to the N-terminus of mouse Casein protein | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P06796 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12994 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.