Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Casein Kinase 1 epsilon Antibody (OTI5D4) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270464
Description
Casein Kinase 1 epsilon Monoclonal specifically detects Casein Kinase 1 epsilon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Casein Kinase 1 epsilon | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
casein kinase 1, epsilon, casein kinase I isoform epsilon, CKIe, CKI-epsilon, EC 2.7.11, EC 2.7.11.1, HCKIE, MGC10398 | |
Mouse | |
47.1 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI5D4 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
CSNK1E | |
Human recombinant protein fragment corresponding to amino acids 153-416 of human CSNK1E (NP_689407) produced in E.coli. | |
Affinity Purified | |
Protein Kinase, Wnt Signaling Pathway | |
1454 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction