Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Casein Kinase 1 gamma Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232245AF750
Description
Casein Kinase 1 gamma Polyclonal antibody specifically detects Casein Kinase 1 gamma in Human, Mouse samples. It is validated for Western BlotSpecifications
Casein Kinase 1 gamma | |
Polyclonal | |
Western Blot | |
casein kinase 1, gamma 1, casein kinase I isoform gamma-1, CKI-gamma 1, EC 2.7.11, EC 2.7.11.1 | |
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Casein Kinase I Isoform gamma-1 using the numbering given in entry NP_071331.2 (GeneID 53944). | |
0.1 mL | |
Protein Kinase | |
53944 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction