Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Casein Kinase 1 gamma Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299829APC
Description
Casein Kinase 1 gamma Polyclonal antibody specifically detects Casein Kinase 1 gamma in Human samples. It is validated for ELISASpecifications
Casein Kinase 1 gamma | |
Polyclonal | |
ELISA | |
casein kinase 1, gamma 1, casein kinase I isoform gamma-1, CKI-gamma 1, EC 2.7.11, EC 2.7.11.1 | |
Produced in rabbits immunized with purified, recombinant Human Casein Kinase 1 gamma (Accession#: NP_071331.2; Met1-Lys422) | |
0.1 mL | |
Protein Kinase | |
53944 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction