Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Casein Kinase 2 alpha Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100378F
Description
Casein Kinase 2 alpha Polyclonal antibody specifically detects Casein Kinase 2 alpha in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Casein Kinase 2 alpha | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
casein kinase 2, alpha 1 polypeptide, casein kinase II alpha 1 subunit, CK II alpha, CK2 catalytic subunit alpha, CK2A1casein kinase II subunit alpha, CKII, EC 2.7.11, EC 2.7.11.1, protein kinase CK2 | |
The immunogen recognized by this antibody maps to a region between residue 350 and the C-terminus (residue 391) of human Casein Kinase 2, alpha 1 using the numbering given in entry NP_001886.1 (GeneID 1457). | |
0.1 mL | |
Cancer, Cell Cycle and Replication, DNA Repair, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
1457 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction