Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Casein Sodium Salt

Catalog No. p-5386198
Click to view available options
Quantity:
500g
1kg
5kg

For research use

α-Casein can be an immunogen in young children, and anti-α-casein antibodies have strong cross-reactivity with insulin. This effect can explain the uncommon insulin autoimmunity, but it does not appear to progress to Type I diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Immunochemistry, Kinase and other Enzyme Assay
Formulation Lyophilized powder
Quantity 1kg
Source Bovine milk
Regulatory Status RUO
Product Type Casein sodium salt
Recombinant Native
Form Lyophilized

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.