Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CASIN

Catalog No. 505050R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-505-0R 50 mg
50-501-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-505-0R Supplier Tocris Bioscience™ Supplier No. 5050/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Cdc42 inhibitor

Cdc42 GTPase inhibitor (IC50 = 2μM). Reduces active levels of Cdc42 in aged hematopoietic stem cells (HSCs) to that in young HSCs. Reverses the aging-related and polarity phenotype of aged HSCs to that of young HSCs in vivo. Increases histone H4K16 acetylation in aged HSCs.

Specifications

Quantity 50 mg
Product Type CASIN
Purity >98%
Molecular Weight (g/mol) 306.4
Inhibitors CASIN

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.