Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CASK Monoclonal Antibody (A1C5)
GREENER_CHOICE

Catalog No. PIMA536121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536121 Supplier Invitrogen™ Supplier No. MA536121
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CASK Monoclonal Antibody for Western Blot, IHC (P)

CASK is a calcium/calmodulin-dependent serine protein kinase which is a MAGUK (membrane-associated guanylate kinase) protein family member. CASK is a scaffold protein and the encoded protein is located at synapses in the brain. CASK associates with FG syndrome 4, mental retardation, microcephaly with pontine and cerebellar hypoplasia, and a form of X-linked mental retardation. CASK functions as a cytoskeletal membrane scaffold that coordinates signal transduction pathways within the cortical cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CASK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A1C5
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Cask
Gene Accession No. O14936, O70589, Q62915
Gene Alias CAGH39; calcium/calmodulin dependent serine protein kinase; calcium/calmodulin-dependent serin protein kinase; Calcium/calmodulin-dependent serine protein kinase; calcium/calmodulin-dependent serine protein kinase (MAGUK family); calcium/calmodulin-dependent serine protein kinase membrane-associated guanylate kinase; CAMGUK; CASK; CMG; DXPri1; DXRib1; FGS4; G-protein-coupled receptor GPR34; hCASK; LIN2; LIN-2; MGC7449; MICPCH; mLin-2; MRXSNA; OTTHUMP00000023157; OTTHUMP00000023161; OTTHUMP00000023162; Pals3; peripheral plasma membrane protein CASK; Protein lin-2 homolog; RP23-278L4.1; TNRC8; trinucleotide repeat containing 8
Gene Symbols Cask
Host Species Mouse
Immunogen Recombinant protein within Human CASK aa 311-513.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12361, 29647, 8573
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.